4-(2-Chlorophenoxy)-3-fluoroaniline

Supplier: Matrix Scientific

022747-500MG
101814-650EA 495.38 USD
101814-650
4-(2-Chlorophenoxy)-3-fluoroaniline
4-(2-Chlorophenoxy)-3-fluoroaniline

Hazard: Irritant

Formula: C₁₂H₉ClFNO
MDL Number: MFCD08687427
CAS Number: 946785-59-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR