4-(2-Methoxyphenoxy)-3-methylaniline

Supplier: Matrix Scientific

022670-500MG
101814-496EA 315.38 USD
101814-496
4-(2-Methoxyphenoxy)-3-methylaniline
4-(2-Methoxyphenoxy)-3-methylaniline

Hazard: Irritant

Formula: C₁₄H₁₅NO₂
MDL Number: MFCD08688077
CAS Number: 946698-59-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR