4-Methoxy-3-methylaniline hydrochloride

Supplier: Matrix Scientific

022656-500MG
101814-468EA 315.38 USD
101814-468
4-Methoxy-3-methylaniline hydrochloride
4-Methoxy-3-methylaniline hydrochloride

Hazard: Irritant

Formula: C₈H₁₂ClNO
MDL Number: MFCD09800634
CAS Number: 1185297-84-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR