4-(3-Ethoxyphenoxy)-2-(trifluoromethyl)aniline

Supplier: Matrix Scientific

022638-500MG
101814-432EA 315.38 USD
101814-432
4-(3-Ethoxyphenoxy)-2-(trifluoromethyl)aniline
4-(3-Ethoxyphenoxy)-2-(trifluoromethyl)aniline

Hazard: Irritant

Formula: C₁₅H₁₄F₃NO₂
MDL Number: MFCD08687752
CAS Number: 946784-84-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR