4-(2-Chloro-5-methylphenoxy)-2-(trifluoromethyl)phenylamine

Supplier: Matrix Scientific

022589-500MG
101814-334EA 315.38 USD
101814-334
4-(2-Chloro-5-methylphenoxy)-2-(trifluoromethyl)phenylamine
4-(2-Chloro-5-methylphenoxy)-2-(trifluoromethyl)phenylamine

Hazard: Irritant

Formula: C₁₄H₁₁ClF₃NO
MDL Number: MFCD08687395
CAS Number: 946663-07-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR