4-[(2-Methylpentyl)oxy]-2-(trifluoromethyl)aniline

Supplier: Matrix Scientific

022578-500MG
101814-312EA 315.38 USD
101814-312
4-[(2-Methylpentyl)oxy]-2-(trifluoromethyl)aniline
4-[(2-Methylpentyl)oxy]-2-(trifluoromethyl)aniline

Hazard: Irritant

Formula: C₁₃H₁₈F₃NO
MDL Number: MFCD08688306
CAS Number: 946741-10-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR