4-Ethoxy-2-(trifluoromethyl)aniline

Supplier: Matrix Scientific

022574-500MG
101814-304EA 315.38 USD
101814-304
4-Ethoxy-2-(trifluoromethyl)aniline
4-Ethoxy-2-(trifluoromethyl)aniline

Hazard: Irritant

Formula: C₉H₁₀F₃NO
MDL Number: MFCD08687760
CAS Number: 121307-27-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR