4-(3-Phenylpropoxy)-2-(trifluoromethyl)aniline

Supplier: Matrix Scientific

022571-500MG
101814-298EA 315.38 USD
101814-298
4-(3-Phenylpropoxy)-2-(trifluoromethyl)aniline
4-(3-Phenylpropoxy)-2-(trifluoromethyl)aniline

Hazard: Irritant

Formula: C₁₆H₁₆F₃NO
MDL Number: MFCD08688485
CAS Number: 946663-01-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR