4-(2-Methoxy-4-methylphenoxy)-3-(trifluoromethyl)phenylamine

Supplier: Matrix Scientific

022547-500MG
101814-252EA 315.38 USD
101814-252
4-(2-Methoxy-4-methylphenoxy)-3-(trifluoromethyl)phenylamine
4-(2-Methoxy-4-methylphenoxy)-3-(trifluoromethyl)phenylamine

Hazard: Irritant

Formula: C₁₅H₁₄F₃NO₂
MDL Number: MFCD08688057
CAS Number: 946784-20-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR