4-(2-Propoxyethoxy)-3-(trifluoromethyl)aniline

Supplier: Matrix Scientific

022505-500MG
101814-168EA 315.38 USD
101814-168
4-(2-Propoxyethoxy)-3-(trifluoromethyl)aniline
4-(2-Propoxyethoxy)-3-(trifluoromethyl)aniline

Hazard: Irritant

Formula: C₁₂H₁₆F₃NO₂
MDL Number: MFCD08688518
CAS Number: 946662-75-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR