4-(1-Naphthylmethoxy)-3-(trifluoromethyl)aniline
Supplier: Matrix Scientific
022496-500MG
101814-150EA
315.38
USD
101814-150
4-(1-Naphthylmethoxy)-3-(trifluoromethyl)aniline
4-(1-Naphthylmethoxy)-3-(trifluoromethyl)aniline
Hazard: Irritant
Formula:
C₁₈H₁₄F₃NO |
MDL Number:
MFCD08688422 CAS Number: 946740-46-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...