N-{2-[4-Amino-2-(trifluoromethyl)phenoxy]ethyl}-N,N-diethylamine

Supplier: Matrix Scientific

022487-500MG
101814-132EA 315.38 USD
101814-132
N-{2-[4-Amino-2-(trifluoromethyl)phenoxy]ethyl}-N,N-diethylamine
N-{2-[4-Amino-2-(trifluoromethyl)phenoxy]ethyl}-N,N-diethylamine

Hazard: Irritant

Formula: C₁₃H₁₉F₃N₂O
MDL Number: MFCD08686838
CAS Number: 689300-09-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR