4-(2,3-Dihydro-1H-inden-5-yloxy)phenylamine

Supplier: Matrix Scientific

022476-500MG
101814-110EA 315.38 USD
101814-110
4-(2,3-Dihydro-1H-inden-5-yloxy)phenylamine
4-(2,3-Dihydro-1H-inden-5-yloxy)phenylamine

Hazard: Irritant

Formula: C₁₅H₁₅NO
MDL Number: MFCD08687595
CAS Number: 937607-41-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR