4-(Cyclohexylmethoxy)aniline

Supplier: Matrix Scientific

022414-500MG
101813-988EA 538.46 USD
101813-988
4-(Cyclohexylmethoxy)aniline
4-(Cyclohexylmethoxy)aniline

Hazard: Irritant

Formula: C₁₃H₁₉NO
MDL Number: MFCD08687495
CAS Number: 76253-28-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR