3-Chloro-4-(2-naphthyloxy)aniline hydrochloride

Supplier: Matrix Scientific

022408-500MG
101813-976EA 315.38 USD
101813-976
3-Chloro-4-(2-naphthyloxy)aniline hydrochloride
3-Chloro-4-(2-naphthyloxy)aniline hydrochloride

Hazard: Irritant

Formula: C₁₆H₁₃Cl₂NO
MDL Number: MFCD01191737
CAS Number: 298220-68-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR