4-[4-(tert-Butyl)phenoxy]-3-chlorophenylamine hydrochloride

Supplier: Matrix Scientific

022396-500MG
101813-952EA 207.69 USD
101813-952
4-[4-(tert-Butyl)phenoxy]-3-chlorophenylamine hydrochloride
4-[4-(tert-Butyl)phenoxy]-3-chlorophenylamine hydrochloride

Hazard: Irritant

Formula: C₁₆H₁₉Cl₂NO
MDL Number: MFCD09800623
CAS Number: 1185296-96-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR