3-Chloro-4-(3-chloro-4-fluorophenoxy)aniline

Supplier: Matrix Scientific

022377-500MG
101813-914EA 315.38 USD
101813-914
3-Chloro-4-(3-chloro-4-fluorophenoxy)aniline
3-Chloro-4-(3-chloro-4-fluorophenoxy)aniline

Hazard: Irritant

Formula: C₁₂H₈Cl₂FNO
MDL Number: MFCD08687231
CAS Number: 946697-16-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR