3-Chloro-4-(2,3-dimethylphenoxy)aniline

Supplier: Matrix Scientific

022369-500MG
101813-898EA 315.38 USD
101813-898
3-Chloro-4-(2,3-dimethylphenoxy)aniline
3-Chloro-4-(2,3-dimethylphenoxy)aniline

Hazard: Irritant

Formula: C₁₄H₁₄ClNO
MDL Number: MFCD07365127
CAS Number: 893754-18-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR