3-Chloro-4-(2-methylphenoxy)aniline

Supplier: Matrix Scientific

022368-500MG
101813-896EA 315.38 USD
101813-896
3-Chloro-4-(2-methylphenoxy)aniline
3-Chloro-4-(2-methylphenoxy)aniline

Hazard: Irritant

Formula: C₁₃H₁₂ClNO
MDL Number: MFCD07365129
CAS Number: 56966-57-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR