3-Chloro-4-(tetrahydro-2-furanylmethoxy)aniline

Supplier: Matrix Scientific

022353-500MG
101813-868EA 315.38 USD
101813-868
3-Chloro-4-(tetrahydro-2-furanylmethoxy)aniline
3-Chloro-4-(tetrahydro-2-furanylmethoxy)aniline

Hazard: Irritant

Formula: C₁₁H₁₄ClNO₂
MDL Number: MFCD08687478
CAS Number: 943117-19-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR