3-Chloro-4-[(2-methylpentyl)oxy]aniline

Supplier: Matrix Scientific

022352-500MG
101813-866EA 315.38 USD
101813-866
3-Chloro-4-[(2-methylpentyl)oxy]aniline
3-Chloro-4-[(2-methylpentyl)oxy]aniline

Hazard: Irritant

Formula: C₁₂H₁₈ClNO
MDL Number: MFCD08687373
CAS Number: 946697-04-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR