4-Butoxy-3-chloroaniline

Supplier: Matrix Scientific

022346-500MG
101813-854EA 315.38 USD
101813-854
4-Butoxy-3-chloroaniline
4-Butoxy-3-chloroaniline

Hazard: Irritant

Formula: C₁₀H₁₄ClNO
MDL Number: MFCD08687150
CAS Number: 5493-75-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR