2-(3-Ethoxyphenoxy)-5-fluoroaniline

Supplier: Matrix Scientific

022333-500MG
101813-828EA 315.38 USD
101813-828
2-(3-Ethoxyphenoxy)-5-fluoroaniline
2-(3-Ethoxyphenoxy)-5-fluoroaniline

Hazard: Irritant

Formula: C₁₄H₁₄FNO₂
MDL Number: MFCD08687719
CAS Number: 946729-58-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR