2-(2,3-Dihydro-1H-inden-5-yloxy)-5-fluorophenylamine

Supplier: Matrix Scientific

022326-500MG
101813-814EA 315.38 USD
101813-814
2-(2,3-Dihydro-1H-inden-5-yloxy)-5-fluorophenylamine
2-(2,3-Dihydro-1H-inden-5-yloxy)-5-fluorophenylamine

Hazard: Irritant

Formula: C₁₅H₁₄FNO
MDL Number: MFCD08687587
CAS Number: 937596-45-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR