2-[4-(sec-Butyl)phenoxy]-5-fluoroaniline

Supplier: Matrix Scientific

022320-500MG
101813-802EA 315.38 USD
101813-802
2-[4-(sec-Butyl)phenoxy]-5-fluoroaniline
2-[4-(sec-Butyl)phenoxy]-5-fluoroaniline

Hazard: Irritant

Formula: C₁₆H₁₈FNO
MDL Number: MFCD08687183
CAS Number: 946684-05-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR