2-Butoxy-5-fluoroaniline

Supplier: Matrix Scientific

022269-500MG
101813-700EA 315.38 USD
101813-700
2-Butoxy-5-fluoroaniline
2-Butoxy-5-fluoroaniline

Hazard: Irritant

Formula: C₁₀H₁₄FNO
MDL Number: MFCD08687151
CAS Number: 946683-76-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR