2-(3-Isopropylphenoxy)-4-methylaniline

Supplier: Matrix Scientific

022226-500MG
101813-614EA 315.38 USD
101813-614
2-(3-Isopropylphenoxy)-4-methylaniline
2-(3-Isopropylphenoxy)-4-methylaniline

Hazard: Irritant

Formula: C₁₆H₁₉NO
MDL Number: MFCD08687987
CAS Number: 946716-45-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR