2-(2,3-Dihydro-1H-inden-5-yloxy)-5-methylphenylamine

Supplier: Matrix Scientific

022168-500MG
101813-498EA 315.38 USD
101813-498
2-(2,3-Dihydro-1H-inden-5-yloxy)-5-methylphenylamine
2-(2,3-Dihydro-1H-inden-5-yloxy)-5-methylphenylamine

Hazard: Irritant

Formula: C₁₆H₁₇NO
MDL Number: MFCD08687590
CAS Number: 946773-95-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR