2-(4-Isopropylphenoxy)-5-methylaniline

Supplier: Matrix Scientific

022161-500MG
101813-484EA 315.38 USD
101813-484
2-(4-Isopropylphenoxy)-5-methylaniline
2-(4-Isopropylphenoxy)-5-methylaniline

Hazard: Irritant

Formula: C₁₆H₁₉NO
MDL Number: MFCD08687990
CAS Number: 946715-98-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR