N-[3-(2-Amino-4-methylphenoxy)phenyl]-N,N-dimethylamine

Supplier: Matrix Scientific

022142-500MG
101813-446EA 315.38 USD
101813-446
N-[3-(2-Amino-4-methylphenoxy)phenyl]-N,N-dimethylamine
N-[3-(2-Amino-4-methylphenoxy)phenyl]-N,N-dimethylamine

Hazard: Irritant

Formula: C₁₅H₁₈N₂O
MDL Number: MFCD08686824
CAS Number: 946728-44-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR