2-(2-Ethoxyethoxy)-5-methylaniline

Supplier: Matrix Scientific

022102-500MG
101813-366EA 495.38 USD
101813-366
2-(2-Ethoxyethoxy)-5-methylaniline
2-(2-Ethoxyethoxy)-5-methylaniline

Hazard: Irritant

Formula: C₁₁H₁₇NO₂
MDL Number: MFCD08687696
CAS Number: 946773-50-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR