2-(Cyclopentylmethoxy)-5-methylaniline

Supplier: Matrix Scientific

022096-500MG
101813-354EA 315.38 USD
101813-354
2-(Cyclopentylmethoxy)-5-methylaniline
2-(Cyclopentylmethoxy)-5-methylaniline

Hazard: Irritant

Formula: C₁₃H₁₉NO
MDL Number: MFCD08687513
CAS Number: 946682-80-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR