Methyl-3-[2-amino-4-(trifluoromethyl)phenoxy]benzoate

Supplier: Matrix Scientific

022065-500MG
101813-292EA 315.38 USD
101813-292
Methyl-3-[2-amino-4-(trifluoromethyl)phenoxy]benzoate
Methyl-3-[2-amino-4-(trifluoromethyl)phenoxy]benzoate

Hazard: Irritant

Formula: C₁₅H₁₂F₃NO₃
MDL Number: MFCD08688151
CAS Number: 946682-72-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR