2-(3-Chloro-4-fluorophenoxy)-5-(trifluoromethyl)phenylamine

Supplier: Matrix Scientific

022061-500MG
101813-284EA 315.38 USD
101813-284
2-(3-Chloro-4-fluorophenoxy)-5-(trifluoromethyl)phenylamine
2-(3-Chloro-4-fluorophenoxy)-5-(trifluoromethyl)phenylamine

Hazard: Irritant

Formula: C₁₃H₈ClF₄NO
MDL Number: MFCD08687332
CAS Number: 946728-05-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR