2-(2-Ethylphenoxy)-5-(trifluoromethyl)aniline

Supplier: Matrix Scientific

022058-500MG
101813-278EA 315.38 USD
101813-278
2-(2-Ethylphenoxy)-5-(trifluoromethyl)aniline
2-(2-Ethylphenoxy)-5-(trifluoromethyl)aniline

Hazard: Irritant

Formula: C₁₅H₁₄F₃NO
MDL Number: MFCD08687789
CAS Number: 946682-70-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR