2-Butoxy-5-(trifluoromethyl)aniline

Supplier: Matrix Scientific

022033-500MG
101813-228EA 315.38 USD
101813-228
2-Butoxy-5-(trifluoromethyl)aniline
2-Butoxy-5-(trifluoromethyl)aniline

Hazard: Irritant

Formula: C₁₁H₁₄F₃NO
MDL Number: MFCD08687162
CAS Number: 121307-24-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR