5-Chloro-2-(3,4-difluorophenoxy)aniline

Supplier: Matrix Scientific

022019-500MG
101813-200EA 315.38 USD
101813-200
5-Chloro-2-(3,4-difluorophenoxy)aniline
5-Chloro-2-(3,4-difluorophenoxy)aniline

Hazard: Irritant

Formula: C₁₂H₈ClF₂NO
MDL Number: MFCD08687263
CAS Number: 946714-90-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR