5-Chloro-2-(4-chloro-3-methylphenoxy)aniline

Supplier: Matrix Scientific

021999-500MG
101813-160EA 315.38 USD
101813-160
5-Chloro-2-(4-chloro-3-methylphenoxy)aniline
5-Chloro-2-(4-chloro-3-methylphenoxy)aniline

Hazard: Irritant

Formula: C₁₃H₁₁Cl₂NO
MDL Number: MFCD00086559
CAS Number: 90207-16-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR