[(5-Chloro-1H-indol-2-yl)methyl]methylamine

Supplier: Matrix Scientific

016184-500MG
101804-260EA 283.08 USD
101804-260
[(5-Chloro-1H-indol-2-yl)methyl]methylamine
[(5-Chloro-1H-indol-2-yl)methyl]methylamine

Hazard: Irritant

Formula: C₁₀H₁₁ClN₂
MDL Number: MFCD05181972
CAS Number: 883528-38-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR