Azepan-1-ylbutanoic acid hydrochloride

Supplier: Matrix Scientific

016171-500MG
101804-236EA 283.08 USD
101804-236
Azepan-1-ylbutanoic acid hydrochloride
Azepan-1-ylbutanoic acid hydrochloride

Hazard: Irritant

Formula: C₁₀H₂₀ClNO₂
MDL Number: MFCD06799773
CAS Number: 54436-99-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR