Piperidin-1-ylbutanoic acid hydrochloride
Supplier: Matrix Scientific
016168-500MG
101804-230EA
192.31
USD
101804-230
Piperidin-1-ylbutanoic acid hydrochloride
Piperidin-1-ylbutanoic acid hydrochloride
Hazard: Irritant
Formula:
C₉H₁₈ClNO₂ Melting Pt: 189…190 °C |
MDL Number:
MFCD06801215 CAS Number: 5463-76-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...