Azepan-1-ylacetic acid hydrochloride

Supplier: Matrix Scientific

016166-500MG
101804-226EA 283.08 USD
101804-226
Azepan-1-ylacetic acid hydrochloride
Azepan-1-ylacetic acid hydrochloride

Hazard: Irritant

Formula: C₈H₁₆ClNO₂
MDL Number: MFCD06799763
CAS Number: 90204-70-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR