[3-(2-Methyl-1H-imidazol-1-yl)propyl]amine dihydrochloride

Supplier: Matrix Scientific

016160-500MG
101804-220EA 283.08 USD
101804-220
[3-(2-Methyl-1H-imidazol-1-yl)propyl]amine dihydrochloride
[3-(2-Methyl-1H-imidazol-1-yl)propyl]amine dihydrochloride

Hazard: Irritant

Formula: C₇H₁₅Cl₂N₃
MDL Number: MFCD06801042
CAS Number: 1059626-17-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR