Cyclopentylhydrazine

Supplier: Matrix Scientific

016146-500MG
101804-192EA 218.46 USD
101804-192
Cyclopentylhydrazine
Cyclopentylhydrazine

Hazard: Irritant/Store in Freezer

Formula: C₅H₁₂N₂
Boiling Pt: 60…65 °C (11 Torr)
MDL Number: MFCD07186351
CAS Number: 30923-92-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR