(4-Methyl-3-fluorobenzyl)hydrazine dihydrochloride

Supplier: Matrix Scientific

016141-500MG
101804-182EA 283.08 USD
101804-182
(4-Methyl-3-fluorobenzyl)hydrazine dihydrochloride
(4-Methyl-3-fluorobenzyl)hydrazine dihydrochloride

Hazard: Irritant

Formula: C₈H₁₃Cl₂FN₂
MDL Number: MFCD06801032
CAS Number: 1185303-71-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR