(3-Methoxy-4-fluorobenzyl)hydrazine dihydrochloride

Supplier: Matrix Scientific

016135-500MG
101804-170EA 283.08 USD
101804-170
(3-Methoxy-4-fluorobenzyl)hydrazine dihydrochloride
(3-Methoxy-4-fluorobenzyl)hydrazine dihydrochloride

Hazard: Irritant

Formula: C₈H₁₃Cl₂FN₂O
MDL Number: MFCD06800925
CAS Number: 1185081-71-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR