Benzylhydrazine dihydrochloride ≥97%
Supplier: Matrix Scientific
016131-5G
016131-25G
101804-160EA
73.85
USD
101804-160
101804-162
Benzylhydrazine dihydrochloride ≥97%
Benzylhydrazine dihydrochloride
Purity: 97%
CAS: 20570-96-1
MDL Number: MFCD00012921
Molecular Formula: C7H12Cl2N2
Molecular Weight: 195,09
Hazard: Toxic
Formula:
C₇H₁₀N₂ Melting Pt: 143…145 °C |
MDL Number:
MFCD00012921 CAS Number: 20570-96-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...