(3-Fluoro-2-methylphenyl)hydrazine hydrochloride

Supplier: Matrix Scientific

016130-500MG
101804-158EA 283.08 USD
101804-158
(3-Fluoro-2-methylphenyl)hydrazine hydrochloride
(3-Fluoro-2-methylphenyl)hydrazine hydrochloride

Hazard: Irritant

Formula: C₇H₁₀ClFN₂
MDL Number: MFCD06800755
CAS Number: 1059626-01-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR