1,3,4,6-Tetrahydropyrrolo[3,2-e][1,4]diazepine-2,5-dione ≥98%

Supplier: Matrix Scientific

016116-250MG
101804-132EA 250.77 USD
101804-132
1,3,4,6-Tetrahydropyrrolo[3,2-e][1,4]diazepine-2,5-dione ≥98%
1,3,4,6-Tetrahydropyrrolo[3,2-e][1,4]diazepine-2,5-dione

Hazard: Irritant

Formula: C₇H₇N₃O₂
MDL Number: MFCD07366981
CAS Number: 1099597-10-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR