1,5-Naphthyridine-2-carboxylic acid ≥98%

Supplier: Matrix Scientific

016106-250MG
101804-106EA 176.92 USD
101804-106
1,5-Naphthyridine-2-carboxylic acid ≥98%
1,5-Naphthyridine-2-carboxylic acid

Hazard: Irritant

Formula: C₉H₆N₂O₂
MDL Number: MFCD06801138
CAS Number: 49850-62-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR